Fittingen, Fingertight
Groen
Catalogus # 554-1151
Leverancier: VICI JOUR RESEARCH
Ga naar:
Product Details & Documents
PEEK one-piece fingertight fittings. Work with all ¹/₁₆'' OD tubings and female 10 to 32 ports. They are rated to 350 bar and can be used with virtually any HPLC fittings, eg. Swagelok, Parker, Rheodyne, Valco.
- 100% inert and biocompatible
- Same colour coding as capillary tubing
Specificaties
- Omschrijving:Groen