Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Fittingen, Fingertight
Fittingen, Fingertight
Blauw
Catalog # 554-1150
Supplier:   VICI JOUR RESEARCH
Fittingen, Fingertight
Catalog # 554-1150
Supplier:  VICI JOUR RESEARCH
Fittingen, Fingertight
Fittingen, Fingertight
Blauw
Price
69,30 €
Log in for availability
1 * 6 ST
Product Details & Documents
PEEK one-piece fingertight fittings. Work with all ¹/₁₆'' OD tubings and female 10 to 32 ports. They are rated to 350 bar and can be used with virtually any HPLC fittings, eg. Swagelok, Parker, Rheodyne, Valco.
  • 100% inert and biocompatible
  • Same colour coding as capillary tubing
Specifications
  • Omschrijving:
    Blauw