Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Fittingen, Fingertight
Fittingen, Fingertight
Zwart
Catalogus # 554-1147
Leverancier:   VICI JOUR RESEARCH
Fittingen, Fingertight
Catalogus # 554-1147
Leverancier:  VICI JOUR RESEARCH
Fittingen, Fingertight
Fittingen, Fingertight
Zwart
Price
69,30 €
Log in voor beschikbaarheid
1 * 6 ST
Product Details & Documents
PEEK one-piece fingertight fittings. Work with all ¹/₁₆'' OD tubings and female 10 to 32 ports. They are rated to 350 bar and can be used with virtually any HPLC fittings, eg. Swagelok, Parker, Rheodyne, Valco.
  • 100% inert and biocompatible
  • Same colour coding as capillary tubing
Specificaties
  • Omschrijving:
    Zwart
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.