Nessler cylinders
Nessler cylinder 50 ml, tall form, with optically plane bottom, circular mark at 50 ml, Ø×L: 24×180 mm
Catalogus # WITG2.150.050
Leverancier: witeg Labortechnik
Ga naar:
Product Details & Documents
Colour comparison cylinders according to Nessler for comparators, with optically plane bottom.
Specificaties
- Omschrijving:Nessler cylinder 50 ml, tall form, with optically plane bottom, circular mark at 50 ml, Ø×L: 24×180 mm
- Seq:01