Orders LinkContactUsLinkComponent
Double spatulas, micro
Double spatulas, micro
150 mm, 6 mm
Catalogus # 231-2322
Leverancier:   RSG
Double spatulas, micro
Catalogus # 231-2322
Leverancier:  RSG
Double spatulas, micro
Double spatulas, micro
150 mm, 6 mm
Price
37,20 €
Log in voor beschikbaarheid
1 * 5 ST
Product Details & Documents
Stainless steel. Anti-magnetic, very flexible micro powder spatula.
  • With round handle, one flat and one scoop shaped, rounded spatula end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
Documents
Specificaties
  • Lengte:
    150 mm
  • Breedte blad:
    6 mm