Double spatulas, powder
Poederspatel, macro, 185 mm, 9 mm
Catalogus # 231-1411
Leverancier: RSG
Ga naar:
Product Details & Documents
Stainless steel.
- With round handle, one flat and one scoop shaped, rounded spatula end
- Straight shape
- Very good thermal resistance, autoclavable
- Good chemical resistance
Specificaties
- Omschrijving:Poederspatel, macro
- Lengte:185 mm
- Breedte blad:9 mm