Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Methyleenchloride, anhydrous ≥99.7% stabilised

Methyleenchloride, anhydrous ≥99.7% stabilised

4x1 L, 4
Catalog # 41835.M1
Health
Irritant
Methyleenchloride, anhydrous ≥99.7% stabilised
Catalog # 41835.M1
CAS Number:  75-09-2
Methyleenchloride, anhydrous ≥99.7% stabilised
Methyleenchloride, anhydrous ≥99.7% stabilised

4x1 L, 4

CAS Number:  75-09-2
Price
375,00 €
Log in for availability
1 * 4 ST
Product Details & Documents

Purity: 99.7+% CAS No.: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93 Physical Form: Liquid Density: 1.325 Refractive Index: 1.4244 Odour: Sweet Melting Point: -95° Boiling Point: 39-40° Beilstein Reference: 1730800 Merck Reference: 14,6063 UN No.: 1593 MDL No.: MFCD00000881

Solvent for anhydrous reactions, water <0.01%.

Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:14,6063
  • Boiling Pt:39…40 °C
  • Melting Pt:–95 °C
  • Density:1,325
  • Storage Temperature:Kamertemperatuur
  • EINECS:200-838-9
  • UN:1593
  • ADR:6.1,III
  •  :Packaged under argon in resealable ChemSeal™ bottles.
  •  :Stabilised with amylene
Specifications

Specifications

  • VPE:
    4
  • Info VPE:
    4x1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.