Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Methyleenchloride, anhydrous ≥99.7% stabilised

Glass bottle, 1 L
Catalog # 41835.K2
Health
Irritant
Methyleenchloride, anhydrous ≥99.7% stabilised
Catalog # 41835.K2
CAS Number:  75-09-2
Methyleenchloride, anhydrous ≥99.7% stabilised
Methyleenchloride, anhydrous ≥99.7% stabilised

Glass bottle, 1 L

CAS Number:  75-09-2
Price
117,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Purity: 99.7+% CAS No.: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93 Physical Form: Liquid Density: 1.325 Refractive Index: 1.4244 Odour: Sweet Melting Point: -95° Boiling Point: 39-40° Beilstein Reference: 1730800 Merck Reference: 14,6063 UN No.: 1593 MDL No.: MFCD00000881

Solvent for anhydrous reactions, water <0.01%.

Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:14,6063
  • Boiling Pt:39…40 °C
  • Melting Pt:–95 °C
  • Density:1,325
  • Storage Temperature:Kamertemperatuur
  • EINECS:200-838-9
  • UN:1593
  • ADR:6.1,III
  •  :Packaged under argon in resealable ChemSeal™ bottles.
  •  :Stabilised with amylene
Specifications

Specifications

  • Verpakking:
    Glass bottle
  • VPE:
    1 L

Specifications