Methyleenchloride, anhydrous ≥99.7% stabilised
Glass bottle, 250 mL
Purity: 99.7+% CAS No.: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93 Physical Form: Liquid Density: 1.325 Refractive Index: 1.4244 Odour: Sweet Melting Point: -95° Boiling Point: 39-40° Beilstein Reference: 1730800 Merck Reference: 14,6063 UN No.: 1593 MDL No.: MFCD00000881
Solvent for anhydrous reactions, water <0.01%.
: Methylene chloride, DCM
- :75-09-2
- :CH₂Cl₂
- :MFCD00000881
- :84.93 g/mol
- :14,6063
- :39…40 °C
- :–95 °C
- :1,325
- :Kamertemperatuur
- :200-838-9
- :1593
- :6.1,III
- :Packaged under argon in resealable ChemSeal™ bottles.
- :Stabilised with amylene
- Verpakking:Glass bottle
- VPE:250 mL



