Hydrometers
Urinometer volgens Vogel, 1,000 - 1,060 g/ml, 0,001 g/ml, 150 mm
Catalog # 216-3189
Supplier: Amarell
Jump to:
Product Details & Documents
Hydrometers specifically designed for urine analyses.
Specifications
- Omschrijving:Urinometer volgens Vogel
- Lengte:150 mm
- Seq:04
- Indeling:0,001 g/ml
- Meetbereik:1,000 - 1,060 g/ml