DETERGENT UNIVERSEEL TICKOPUR TR 7 1 * 1 L
Catalogus # 142-1980
Leverancier: BANDELIN-ELECTRONIC
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
Leverancier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Leverancier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Leverancier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
VWR®, Nitrilhandschoenen
Leverancier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Natriumchloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Leverancier: Merck
Ethanol, GPR RECTAPUR®
Leverancier: VWR Chemicals
Cleanroom Waste Receptacles
Leverancier: Palbam