Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Hafnium dioxide, tech.

Hafnium dioxide, tech.

Glass bottle for solids, 500 g
Catalog # 40270.36
Hafnium dioxide, tech.
Catalog # 40270.36
CAS Number:  12055-23-1
Hafnium dioxide, tech.
Hafnium dioxide, tech.

Glass bottle for solids, 500 g

CAS Number:  12055-23-1
Price
998,00 €
Log in for availability
1 * 500 g
Product Details & Documents

CAS No.: 12055-23-1 Molecular Formula: HfO2 Formula Weight: 210.49 Physical Form: -325 Mesh Powder Density: 9.68 Odour: Odourless Merck Reference: 14,4588 MDL No.: MFCD00003565

  • CAS Number:12055-23-1
  • Formula:HfO₂
  • MDL Number:MFCD00003565
  • Molecular Weight:210.49 g/mol
  • Merck Index:14,4588
  • Boiling Pt:5400 °C (1013 hPa)
  • Melting Pt:2774 °C
  • Density:9,68
  •  :Density: 9.68 g/cm³ (20 °C)
  • Storage Temperature:Kamertemperatuur
  • EINECS:235-013-2
Specifications

Specifications

  • Verpakking:
    Glass bottle for solids
  • VPE:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.