Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Hafnium dioxide, tech.

Hafnium dioxide, tech.

Glass bottle for solids, 100 g
Catalogus # 40270.22
Leverancier:   Thermo Fisher Scientific
Hafnium dioxide, tech.
Catalogus # 40270.22
CAS-nummer:  12055-23-1
Hafnium dioxide, tech.
Hafnium dioxide, tech.

Glass bottle for solids, 100 g

CAS-nummer:  12055-23-1
Price
231,00 €
Log in voor beschikbaarheid
1 * 100 g
Product Details & Documents

CAS No.: 12055-23-1 Molecular Formula: HfO2 Formula Weight: 210.49 Physical Form: -325 Mesh Powder Density: 9.68 Odour: Odourless Merck Reference: 14,4588 MDL No.: MFCD00003565

  • CAS-nummer:12055-23-1
  • Formule:HfO₂
  • MDL-nummer:MFCD00003565
  • Molecular Weight:210.49 g/mol
  • Merck index:14,4588
  • Kokend Pt:5400 °C (1013 hPa)
  • Smelten Pt:2774 °C
  • Dichtheid:9,68
  •  :Density: 9.68 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:235-013-2
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    100 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.