Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Lead ≥99.5%, granules

Lead ≥99.5%, granules

Glass bottle for solids, 2 kg
Catalog # 36227.A3
Health
Irritant
Environment
Lead ≥99.5%, granules
Catalog # 36227.A3
CAS Number:  7439-92-1
Lead ≥99.5%, granules
Lead ≥99.5%, granules

Glass bottle for solids, 2 kg

CAS Number:  7439-92-1
Price
613,00 €
Log in for availability
1 * 2 kg
Product Details & Documents

Purity: 99.5% min CAS No.: 7439-92-1 Appearance: Grey Odour: Odourless MDL No.: MFCD00134050

  • CAS Number:7439-92-1
  • Formula:Pb
  • MDL Number:MFCD00134050
  • Molecular Weight:207.2 g/mol
  • Merck Index:12,05410
  • Boiling Pt:1740 °C (1013 hPa)
  • Melting Pt:327 °C
  • Density:11,3437 g/cm³ (20 °C)
  • Storage Temperature:Kamertemperatuur
  • EINECS:231-100-4
Specifications

Specifications

  • Verpakking:
    Glass bottle for solids
  • VPE:
    2 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.