Salpeterzuur standardised solution 1.0 N
Glass bottle, 500 mL
Leverancier: Thermo Fisher Scientific
Salpeterzuur standardised solution 1.0 N
Catalogus # 35624.AP
Leverancier: Thermo Fisher Scientific
CAS-nummer: 7697-37-2
Ga naar:
Product Details & Documents
CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Appearance: Colourless UN No.: 3264 MDL No.: MFCD00011349
Synoniemen: Nitric acid solution
- CAS-nummer:7697-37-2
- Formule:HNO₃
- MDL-nummer:MFCD00011349
- Molecular Weight:63.01 g/mol
- Dichtheid:1,035 g/cm³ (25 °C)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:231-714-2
- UN:3264
- ADR:8,III
- :Density: 1,51 g/cm³ (20 °C)
Documents
Specificaties
Specificaties
- Verpakking:Glass bottle
- VPE:500 mL



