Orders LinkContactUsLinkComponent
Rods, ceramic, round, solid, AL23
Rods, ceramic, round, solid, AL23
6,0 mm, 200 mm
N° de catalogue 32933.DR
Fournisseur:   Thermo Fisher Scientific
Rods, ceramic, round, solid, AL23
N° de catalogue 32933.DR
Rods, ceramic, round, solid, AL23
6,0 mm, 200 mm
Prix
60,40 €
Log in for availability
1 * 200 mm
Product Details & Documents
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Specifications
  • Ø uitw.:
    6,0 mm
  • Lengte:
    200 mm
  • Seq:
    49