Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Bromophenol blue ACS

Glass bottle for solids, 5 g
Catalogus # 32641.06
Leverancier:   Thermo Fisher Scientific
Bromophenol blue ACS
Catalogus # 32641.06
CAS-nummer:  115-39-9
Bromophenol blue ACS
Bromophenol blue ACS

Glass bottle for solids, 5 g

CAS-nummer:  115-39-9
Price
44,60 €
Log in voor beschikbaarheid
1 * 5 g
Product Details & Documents

CAS No.: 115-39-9 Molecular Formula: C19H10Br4O5S Formula Weight: 669.98 Physical Form: Crystalline Odour: Odourless Melting Point: 270° dec. Beilstein Reference: 61698 Merck Reference: 14,1444 MDL No.: MFCD00005875

Synoniemen:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS-nummer:115-39-9
  • Formule:C₁₉H₁₀Br₄O₅S
  • MDL-nummer:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck index:14,1444
  • Smelten Pt:270 °C (ontbinding)
  • Dichtheid:1 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:204-086-2
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    5 g

Specificaties