Basic Violet 3 90+% ACS
Glass bottle for solids, 100 g
Leverancier: Thermo Fisher Scientific
Basic Violet 3 90+% ACS
Catalogus # 22866.22
Leverancier: Thermo Fisher Scientific
CAS-nummer: 548-62-9
Ga naar:
Product Details & Documents
Purity: 90+% CAS No.: 548-62-9 Molecular Formula: C25H30ClN3 Formula Weight: 407.99 Physical Form: Powder Melting Point: ca 215° dec. Beilstein Reference: 4077708 Merck Reference: 14,4395 UN No.: 3077 MDL No.: MFCD00011750
Synoniemen: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
- CAS-nummer:548-62-9
- Formule:C₂₅H₃₀ClN₃
- MDL-nummer:MFCD00011750
- Molecular Weight:407.99 g/mol
- Merck index:14,4395
- Smelten Pt:∼ 215 °C (ontbinding)
- Dichtheid:1,19 g/cm³ (20 °C)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:208-953-6
- UN:3077
- ADR:9,III
Documents
Certificaten
Zoeken op lotnummerSpecificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:100 g
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



