Styreen ≥99.5% stabilised
Plastic bottle, 250 mL
Leverancier: Thermo Fisher Scientific
Styreen ≥99.5% stabilised
Catalogus # 16199.AK
Leverancier: Thermo Fisher Scientific
CAS-nummer: 100-42-5
Ga naar:
Product Details & Documents
Stabilized with 4-tert-butylcatechol
Purity: 99.5% CAS No.: 100-42-5 Molecular Formula: C8H8 Formula Weight: 104.15 Storage Temperature: 2°C to 8°C Flash Point: 31°(88°F) Physical Form: Liquid Density: 0.906 Refractive Index: 1.5460 Sensitivity: Light Sensitive Appearance: Colourless Odour: Pungent pH: neutral Melting Point: -31° Boiling Point: 145-146° Beilstein Reference: 1071236 Merck Reference: 14,8860 UN No.: 2055 MDL No.: MFCD00008612
Synoniemen: Phenylethene, Phenylethylene, Vinylbenzene
- CAS-nummer:100-42-5
- Formule:C₈H₈
- MDL-nummer:MFCD00008612
- Molecular Weight:104.15 g/mol
- Merck index:14,8860
- Kokend Pt:145…146 °C
- Smelten Pt:–31 °C
- Dichtheid:0,906
- Flits Pt:31 °C (88 °F)
- Temperatuur bij opslag:Koelkast
- UN:2055
- ADR:3,III
- :Stabilised with 4-tert-butylpyrocatechol (TBC)
- EINECS:202-851-5
Documents
Specificaties
Specificaties
- Verpakking:Plastic bottle
- VPE:250 mL



