Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Copper(I) bromide ≥98%

Copper(I) bromide ≥98%

1 kg
Catalogus # 11605.A1
Irriterend
Leverancier:   Thermo Fisher Scientific
Copper(I) bromide ≥98%
Catalogus # 11605.A1
CAS-nummer:  7787-70-4
Copper(I) bromide ≥98%
Copper(I) bromide ≥98%

1 kg

CAS-nummer:  7787-70-4
Price
209,00 €
Log in voor beschikbaarheid
1 * 1 kg
Product Details & Documents

Purity: 98% CAS No.: 7787-70-4 Molecular Formula: BrCu Formula Weight: 143.45 Physical Form: Powder Density: 4.98 Appearance: Pale green Melting Point: 492° Boiling Point: 1345° Solubility: Slightly soluble in cold water. Decomposes in hot water. Soluble in HBr, HCl, NH4OH Merck Reference: 14,2659 UN No.: 3077 MDL No.: MFCD00010969

Catalyst

  • CAS-nummer:7787-70-4
  • Formule:BrCu
  • MDL-nummer:MFCD00010969
  • Molecular Weight:143.45 g/mol
  • Merck index:14,2659
  • Kokend Pt:1345 °C
  • Smelten Pt:492 °C
  • Dichtheid:4,98
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:232-131-6
  • ADR:9,III
  •  :Density: 4,71 g/cm³ (20 °C)
Specificaties

Specificaties

  • VPE:
    1 kg

Specificaties