Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Silver cyanide ≥99%

25 g
Catalogus # 11424.14
Giftig
Corrosief
Leverancier:   Thermo Fisher Scientific
Silver cyanide ≥99%
Catalogus # 11424.14
CAS-nummer:  506-64-9
Silver cyanide ≥99%
Silver cyanide ≥99%

25 g

CAS-nummer:  506-64-9
Price
176,00 €
Log in voor beschikbaarheid
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 506-64-9 Molecular Formula: CAgN Formula Weight: 133.84 Physical Form: Powder Density: 3.95 Refractive Index: 1.685 Sensitivity: Light Sensitive Appearance: White to off-white Odour: Odourless Melting Point: 320° dec. Solubility: Insoluble in water, dilute acids, and alcohol. Soluble in alkali cyanides and boiling concentrated HNO3 Merck Reference: 14,8512 UN No.: 1684 MDL No.: MFCD00003409

Silver plating

Synoniemen:  Argentous cyanide
  • CAS-nummer:506-64-9
  • Formule:AgCN
  • MDL-nummer:MFCD00003409
  • Molecular Weight:133.89 g/mol
  • Merck index:14,8512
  • Smelten Pt:320 °C (ontbinding)
  • Dichtheid:3,95
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:208-048-6
  • UN:1684
  • ADR:6.1,II
  •  :Density: 3.943 g/cm³ (20 °C)
Specificaties

Specificaties

  • VPE:
    25 g

Specificaties