Orders LinkContactUsLinkComponent
Silica standard solution
Silica standard solution
Silica standard solution, 0,5 mg/l as SiO2 (NIST), 3,78 L
Catalogus # HACH2100817
Leverancier:   Hach
Silica standard solution
Catalogus # HACH2100817
Leverancier:  Hach
CAS-nummer:  
Silica standard solution
Silica standard solution, 0,5 mg/l as SiO2 (NIST), 3,78 L
CAS-nummer:  
Price
121,00 €
Log in voor beschikbaarheid
1 * 3,78 L
Product Details & Documents
Standard solutions for accuracy checks in silica determinations.
Specificaties
Specificaties
  • Concentratie:
    0,5 mg/l as SiO<sub>2 </sub> (NIST)
  • VPE:
    3,78 L
  • Omschrijving:
    Silica standard solution
Specificaties