Xyleen, BAKER, J.T.Baker®
Glass bottle, 2,5 L
Leverancier: Avantor
Ga naar:
Product Details & Documents
Synoniemen: Dimethylbenzene
- CAS-nummer:1330-20-7
- Formule:C₆H₄(CH₃)₂
- MDL-nummer:MFCD00077264
- Molecular Weight:106.17 g/mol
- Merck index:13,10136
- Kokend Pt:140 °C (1013 hPa)
- Smelten Pt:–34 °C
- Dichtheid:0,86…0.88 g/cm³ (20 °C)
- Flits Pt:25 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:215-535-7
- UN:1307
- ADR:3,III
Documents
Specificaties
Specificatie testresultaten
- Appearance:passes test
- Additional Specification(s)::-
- Assay:min. 98 %
- Color:max. 10
Specificaties
- Verpakking:Glass bottle
- VPE:2,5 L
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



