Silver cyanide ≥99%
100 g
Purity: 99% CAS No.: 506-64-9 Molecular Formula: CAgN Formula Weight: 133.84 Physical Form: Powder Density: 3.95 Refractive Index: 1.685 Sensitivity: Light Sensitive Appearance: White to off-white Odour: Odourless Melting Point: 320° dec. Solubility: Insoluble in water, dilute acids, and alcohol. Soluble in alkali cyanides and boiling concentrated HNO3 Merck Reference: 14,8512 UN No.: 1684 MDL No.: MFCD00003409
Silver plating
: Argentous cyanide
- :506-64-9
- :AgCN
- :MFCD00003409
- :133.89 g/mol
- :14,8512
- :320 °C (ontbinding)
- :3,95
- :Kamertemperatuur
- :208-048-6
- :1684
- :6.1,II
- :Density: 3.943 g/cm³ (20 °C)
- VPE:100 g