Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Silver cyanide ≥99%
100 g
  11424.22
Silver cyanide ≥99%
  11424.22
 :  506-64-9
Silver cyanide ≥99%
Silver cyanide ≥99%
100 g
 :  506-64-9
517,00 €
1 * 100 g
Purity: 99% CAS No.: 506-64-9 Molecular Formula: CAgN Formula Weight: 133.84 Physical Form: Powder Density: 3.95 Refractive Index: 1.685 Sensitivity: Light Sensitive Appearance: White to off-white Odour: Odourless Melting Point: 320° dec. Solubility: Insoluble in water, dilute acids, and alcohol. Soluble in alkali cyanides and boiling concentrated HNO3 Merck Reference: 14,8512 UN No.: 1684 MDL No.: MFCD00003409
Silver plating
 :  Argentous cyanide
  •  :506-64-9
  •  :AgCN
  •  :MFCD00003409
  •  :133.89 g/mol
  •  :14,8512
  •  :320 °C (ontbinding)
  •  :3,95
  •  :Kamertemperatuur
  •  :208-048-6
  •  :1684
  •  :6.1,II
  •  :Density: 3.943 g/cm³ (20 °C)
  • VPE:
    100 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.