Orders LinkContactUsLinkComponent
Microtubes, with Thread
Microtubes, with Thread
Micro tubes with neutral screw cap assembled, sterile, with skirted base, 2,0 ml, 1000/case
Catalogus # SARS72.694.005
Marketsource
Leverancier:   SARSTEDT
Microtubes, with Thread
Catalogus # SARS72.694.005
Leverancier:  SARSTEDT
Marketsource
Microtubes, with Thread
Micro tubes with neutral screw cap assembled, sterile, with skirted base, 2,0 ml, 1000/case
Price
129,41 €
Log in voor beschikbaarheid
1 * 1.000 ST
Product Details & Documents
PP, translucent.
  • Safe screw cap with O-ring
  • Sterile or non sterile
  • Autoclavable
For transport, storage and sample processing in the laboratory.
Specificaties
  • Verpakt:
    1000/case
  • Capaciteit:
    2,0 ml
  • Omschrijving:
    Micro tubes with neutral screw cap assembled, sterile, with skirted base
  • Seq:
    21