Microtubes, Prepared
Microtubes, prepared, glucose fluoride, with attached push cap closure, 1,3 ml, 1000/case
Catalogus # SARS41.1505.005
Marketsource
Leverancier: SARSTEDT
Ga naar:
Product Details & Documents
PP, translucent with PP screw cap.
- Type of preparation: Trisodium citrate solution
- With paper label
- Graduated
- Recommended centrifugation: 1800×g
With screw cap or push-on cap.
Specificaties
- Verpakt:1000/case
- Capaciteit:1,3 ml
- Omschrijving:Microtubes, prepared, glucose fluoride, with attached push cap closure
- Seq:8