Support rings
Zware steunring, met klem, open, met rubberen coating, 70 mm
Catalogus # 241-0327
Leverancier: RSG
Ga naar:
Product Details & Documents
These heavy-duty support rings are made from stainless steel.
- 8 mm rod
- Available with rubber covered rings
Specificaties
- Ø inw.:70 mm
- Omschrijving:Zware steunring, met klem, open, met rubberen coating