Support rings
Zware steunring, met staaf, open, met rubberen coating, 130 mm
Catalog # 241-0326
Supplier: RSG
Jump to:
Product Details & Documents
These heavy-duty support rings are made from stainless steel.
- 8 mm rod
- Available with rubber covered rings
Specifications
- Ø inw.:130 mm
- Omschrijving:Zware steunring, met staaf, open, met rubberen coating