Dubbele statiefkoppeling
Bosshead without blue plastic headed screw, Cast legering, 16 mm
Catalogus # RLAA2005/1PL
Leverancier: R L ENTERPRISES
Ga naar:
Product Details & Documents
Bossheads made from nickel plated cast alloy.
- Either with or without blue plastic headed nickel plated screws
- For use with rods up to 16 mm
Specificaties
- Omschrijving:Bosshead without blue plastic headed screw
- Materiaal:Cast legering
- Voor staaf Ø:16 mm