Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Pierce™ Disposable Columns
Pierce™ Disposable Columns
Disposable column trial pack, includes accessories plus 2× each Cat. No's 29920, 29922 and 29924, and 1× Cat. No. 29923
Catalogus # PIER29925
Leverancier:   Thermo Fisher Scientific
Pierce™ Disposable Columns
Catalogus # PIER29925
Pierce™ Disposable Columns
Pierce™ Disposable Columns
Disposable column trial pack, includes accessories plus 2× each Cat. No's 29920, 29922 and 29924, and 1× Cat. No. 29923
Price
131,00 €
Log in voor beschikbaarheid
1 * 7 ST
Product Details & Documents
Empty polystyrene columns with matching frits and caps for use in a variety of resin based affinity or desalting techniques.
  • For benchtop gravity flow affinity purification and size exclusion chromatography
  • Supplied complete with porous polyethylene discs, stoppers and end caps
  • Compatible with most types of aqueous buffer eluents
  • Plastic funnels can be attached to expand the buffer reservoir for increased flow rates
Specificaties
  • Omschrijving:
    Disposable column trial pack, includes accessories plus 2× each Cat. No's 29920, 29922 and 29924, and 1× Cat. No. 29923
  • Seq:
    04
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Disposable PP funnels (buffer reservoirs) that fit Pierce™ disposable columns
    Disposable PP funnels (buffer reservoirs) that fit Pierce™ disposable columns
    PIER29923
    Log in voor beschikbaarheid
    1 * 50 ST - 219,00 €