Pierce™ Disposable Columns
Disposable PS columns, for packing 1 - 5 ml bed volumes
Catalogus # PIER29922
Leverancier: Thermo Fisher Scientific
Ga naar:
Product Details & Documents
Empty polystyrene columns with matching frits and caps for use in a variety of resin based affinity or desalting techniques.
- For benchtop gravity flow affinity purification and size exclusion chromatography
- Supplied complete with porous polyethylene discs, stoppers and end caps
- Compatible with most types of aqueous buffer eluents
- Plastic funnels can be attached to expand the buffer reservoir for increased flow rates
Specificaties
- Omschrijving:Disposable PS columns, for packing 1 - 5 ml bed volumes
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Disposable PP funnels (buffer reservoirs) that fit Pierce™ disposable columnsDisposable PP funnels (buffer reservoirs) that fit Pierce™ disposable columnsPIER29923Log in voor beschikbaarheid1 * 50 ST - 219,00 €