Orders LinkContactUsLinkComponent
Pierce™ Disposable Columns
Pierce™ Disposable Columns
Disposable PS columns, for packing 0,5 - 2,0 ml bed volumes
Catalogus # PIER29920
Leverancier:   Thermo Fisher Scientific
Pierce™ Disposable Columns
Catalogus # PIER29920
Pierce™ Disposable Columns
Pierce™ Disposable Columns
Disposable PS columns, for packing 0,5 - 2,0 ml bed volumes
Price
381,00 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents
Empty polystyrene columns with matching frits and caps for use in a variety of resin based affinity or desalting techniques.
  • For benchtop gravity flow affinity purification and size exclusion chromatography
  • Supplied complete with porous polyethylene discs, stoppers and end caps
  • Compatible with most types of aqueous buffer eluents
  • Plastic funnels can be attached to expand the buffer reservoir for increased flow rates
Specificaties
  • Omschrijving:
    Disposable PS columns, for packing 0,5 - 2,0 ml bed volumes
  • Seq:
    01
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Disposable PP funnels (buffer reservoirs) that fit Pierce™ disposable columns
    Disposable PP funnels (buffer reservoirs) that fit Pierce™ disposable columns
    PIER29923
    Log in voor beschikbaarheid
    1 * 50 ST - 219,00 €