Accessories for face shields
Replacement visor lens, Celluloseacetaat
Catalogus # 111-2543
Leverancier: UVEX
Ga naar:
Product Details & Documents
Specificaties
- Omschrijving:Replacement visor lens
- Materiaal:Celluloseacetaat
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.