Uitdampschalen
25 ml, 15 mm, 79 mm
Catalogus # 216-0293
Leverancier: TECHNICAL QUARTZ SOLUTIONS
Ga naar:
Product Details & Documents
Made of silica.
- Very good chemical resistance
- Very good temperature resistance
- Easy to clean
Specificaties
- Ø uitw.:79 mm
- Capaciteit:25 ml
- Hoogte:15 mm