Closures for headspace vials
Red PTFE coated butyl rubber cap, pre-assembled
Catalogus # PERCB4000025
Leverancier: PerkinElmer
Ga naar:
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Specificaties
- Omschrijving:Red PTFE coated butyl rubber cap, pre-assembled
- Seq:07