Nessler reagent
30 mL
Fournisseur: Lovibond
Aller à :
Product Details & Documents
Synonymes: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution
- Numéro CAS:7783-33-7
- Numéro MDL:MFCD00049656
- Point d'ébullition:∼100 °C (1013 hPa)
- Densité:1,2 g/cm³ (20 °C)
- UN:2922
- ADR:8,II
Documents
Certificats
Rechercher par numéro de lotSpécifications
Spécifications
- VPE:30 mL
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



