Accessories for comparators
Phosphate, 3/12, 0 - 80 mg/l
Catalogus # LOVI231200
Leverancier: Lovibond
Ga naar:
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
- Disc codes starting with 'N' are for use with the Nessleriser 2150
- Disc codes starting with 'C' are for use with the Nessleriser 2250
- All other discs are for use with the Comparator 2000+
Specificaties
- Meetbereik:0 - 80 mg/l
- Seq:67
- Code:3/12
- Testmethode:Phosphate