Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Accessories for comparators
Phosphate, 3/12, 0 - 80 mg/l
Catalogus # LOVI231200
Leverancier:   Lovibond
Accessories for comparators
Catalogus # LOVI231200
Leverancier:  Lovibond
Accessories for comparators
Accessories for comparators
Phosphate, 3/12, 0 - 80 mg/l
Price
263,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+
Specificaties
  • Meetbereik:
    0 - 80 mg/l
  • Seq:
    67
  • Code:
    3/12
  • Testmethode:
    Phosphate