SURE competent cells
SURE 2 supercompetent cells kit, High-efficiency derivative for cloning DNA with secondary structures, >1×10⁹ transformants/µg, 1 Kit
Catalogus # STRA200152
Marketsource
Leverancier: STRATAGENE
Ga naar:
Product Details & Documents
SURE 2 competent cells are ideal for routine cloning of DNA with secondary structures.
- Lack components of the pathways that catalyse the rearrangement and deletion of nonstandard secondary and tertiary structures
- Allows for blue-white colour screening
Replicating eukaryotic DNA in prokaryotic cells can be problematic. Particular eukaryotic genes may contain inverted repeats or secondary structures, such as Z-DNA, that can be rearranged or deleted by E. coli DNA repair systems.
SURE competent cells are deficient in the E. coli genes involved in the rearrangement and deletion of DNA, thus improving cloning efficiencies of DNA containing irregular structures in prokaryotic cells. SURE cells are restriction minus (McrA-, McrCB-, McrF-, Mrr-, HsdR-) endonuclease (endA) deficient, and recombination (recB recJ) deficient.
Let op: For research use only. Not for use in diagnostic procedures.
Specificaties
- Omschrijving:SURE 2 supercompetent cells kit
- Seq:07
- Info VPE:1 Kit
- Eigenschap:High-efficiency derivative for cloning DNA with secondary structures, >1×10⁹ transformants/µg