Fractogel® EMD DEAE (S) zwakke anionwisselaar, suspensie in 20% ethanol en 150 mM NaCl (20-40 µm) 1 * 10 ml
Catalogus # 1.16888.0010
Leverancier: MERCK PRODUCTION CHEMICALS
Fractogel® EMD DEAE (S) zwakke anionwisselaar, suspensie in 20% ethanol en 150 mM NaCl (20-40 µm) 1 * 10 ml
Catalogus # 1.16888.0010
Leverancier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Leverancier: HEATHROW
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
VWR®, Nitrilhandschoenen
Leverancier: VWR Collection
VWR®, gezichtstissues
Leverancier: VWR Collection
pH-teststrips, MQuant®, Supelco®
Leverancier: Merck
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
VWR®, Transferpipetten, Hoge Prestaties
Leverancier: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschoenen
Leverancier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
VWR®, Serologische pipetten, standaardreeks
Leverancier: VWR Collection
Nitril handschoenen, Kimtech™ Purple Nitrile™
Leverancier: KIMBERLY CLARK