ContactUsLinkComponent
SGE SYRINGE 100µL REMOVABLE NEEDLE GAS TIGHT PLUNGER FOR VARIAN 8100& 8200 NEEDLE IS STAINLESS STEEL 0.50 MM OD (25 GAUGE) 53 MM LON G WITH SIDE HOLE POINT STYLE.
Catalogus # 549-0401
SGE SYRINGE 100µL REMOVABLE NEEDLE GAS TIGHT PLUNGER FOR VARIAN 8100& 8200 NEEDLE IS STAINLESS STEEL 0.50 MM OD (25 GAUGE) 53 MM LON G WITH SIDE HOLE POINT STYLE.
Catalogus # 549-0401
Frequently Bought Together