Order Entry
Belgium
ContactUsLinkComponent
SGE SYRINGE 100µL REMOVABLE NEEDLE NEEDLE IS STAINLESS STEEL 42 MM LONG 23 GAUGE (0.63 MM OD) WITH A CONE POINT STYLE. FOR MANUAL US E ON AGILENT SYSTEMS.
Catalogus # 549-0389
SGE SYRINGE 100µL REMOVABLE NEEDLE NEEDLE IS STAINLESS STEEL 42 MM LONG 23 GAUGE (0.63 MM OD) WITH A CONE POINT STYLE. FOR MANUAL US E ON AGILENT SYSTEMS.
Catalogus # 549-0389
Frequently Bought Together