ContactUsLinkComponent
SGE SYRINGE 100µL REMOVABLE NEEDLE NEEDLE IS STAINLESS STEEL 51 MM LONG 22 GAUGE (0.71 MM OD) WITH LC POINT STYLE AND FITTED WITH A REPEATING ADAPTER
Catalogus # 549-0469
SGE SYRINGE 100µL REMOVABLE NEEDLE NEEDLE IS STAINLESS STEEL 51 MM LONG 22 GAUGE (0.71 MM OD) WITH LC POINT STYLE AND FITTED WITH A REPEATING ADAPTER
Catalogus # 549-0469
Frequently Bought Together