SGE SYRINGE 25µL REMOVABLE NEEDLE IS STAINLESS STEEL 0.50 MM OD (25 GAUGE) 50 MM LONG WITH BEVEL POINT STYLE. REAR FLANGE IS THREADE D AND HAS A PTFE SEAL FOR A GAS TIGHT SEAL WITH REPEATING ADAPTER
Catalogus # SGEA003610
Leverancier: Trajan Scientific and Medical
Frequently Bought Together