Order Entry
Belgium
ContactUsLinkComponent
SGE SYRINGE 25µL REMOVABLE NEEDLE IS 0.71 MM OD (22 GAUGE) 51 MM LONG AND HAS A LC POINT STYLE. FITTED WITH A REPEATING ADAPTER
Catalogus # 549-0467
SGE SYRINGE 25µL REMOVABLE NEEDLE IS 0.71 MM OD (22 GAUGE) 51 MM LONG AND HAS A LC POINT STYLE. FITTED WITH A REPEATING ADAPTER
Catalogus # 549-0467
Frequently Bought Together