ContactUsLinkComponent
SGE SYRINGE 25µL REMOVABLE NEEDLE IS 0.71 MM OD (22 GAUGE) 51 MM LONG AND HAS LC POINT STYLE
Catalogus # 549-0457
SGE SYRINGE 25µL REMOVABLE NEEDLE IS 0.71 MM OD (22 GAUGE) 51 MM LONG AND HAS LC POINT STYLE
Catalogus # 549-0457
Frequently Bought Together