Jump to:
Product Details & Documents
Synonyms: 3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one
- CAS Number:596-01-0
- Formula:C₂₈H₁₈O₄
- MDL Number:MFCD00036202
- Molecular Weight:418.45 g/mol
- :M-Clarity™ quality level = MQ200
- EINECS:209-875-5
Documents
msds_106246
SDS
Specifications
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:253 - 255 °C
- Molar Mass:418.45 g/mol
- Bulk Density:350 kg/m3
Specifications
- Verpakking:Glass bottle
- VPE:5 g



