Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
α-Naphtholphthalein

α-Naphtholphthalein

Glass bottle, 1 g
Catalogus # 1.06246.0001
Leverancier:   Merck
α-Naphtholphthalein
Catalogus # 1.06246.0001
Leverancier:  Merck
CAS-nummer:  596-01-0
α-Naphtholphthalein

Glass bottle, 1 g

CAS-nummer:  596-01-0
Price
377,00 €
Log in voor beschikbaarheid
1 * 1 g
Product Details & Documents
Synoniemen:  3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one
  • CAS-nummer:596-01-0
  • Formule:C₂₈H₁₈O₄
  • MDL-nummer:MFCD00036202
  • Molecular Weight:418.45 g/mol
  •  :M-Clarity™ quality level = MQ200
  • EINECS:209-875-5

Documents

Specificaties

Specificatie testresultaten

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    253 - 255 °C
  • Molar Mass:
    418.45 g/mol
  • Bulk Density:
    350 kg/m3

Specificaties

  • Verpakking:
    Glass bottle
  • VPE:
    1 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.