SGE SYRINGE 10µL REMOVABLE NEEDLE THAT IS 0.47 MM OD (26 GAUGE) 70 MM LONG W/ BEVEL POINT STYLE
Catalogus # 549-0282
Leverancier: Trajan Scientific and Medical
SGE SYRINGE 10µL REMOVABLE NEEDLE THAT IS 0.47 MM OD (26 GAUGE) 70 MM LONG W/ BEVEL POINT STYLE
Catalogus # 549-0282
Leverancier: Trajan Scientific and Medical
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Leverancier: HEATHROW
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
VWR®, Transferpipetten, Hoge Prestaties
Leverancier: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschoenen
Leverancier: VWR Collection
VWR®, Serologische pipetten, standaardreeks
Leverancier: VWR Collection
Nitril handschoenen, Kimtech™ Purple Nitrile™
Leverancier: KIMBERLY CLARK
VWR®, Nitrilhandschoenen
Leverancier: VWR Collection
VWR®, gezichtstissues
Leverancier: VWR Collection
pH-teststrips, MQuant®, Supelco®
Leverancier: Merck
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals