SGE SYRINGE 5µL REMOVABLE NEEDLE NEEDLE IS STAINLESS STEEL 50 MM LONG 26 GAUGE (0.47 MM OD) WITH A BEVEL POINT STYLE. PLUNGER IS MAD E OF NICKEL/TITANIUM ALLOY THAT WILL NOT PERMANENTLY BEND
Catalogus # 549-0271
Leverancier: Trajan Scientific and Medical
Frequently Bought Together