Orders LinkContactUsLinkComponent
Base materials
Base materials
Peptone from meat, 250 g
Catalogus # SERA48620.04
Base materials
Catalogus # SERA48620.04
Leverancier:  
CAS-nummer:  
Base materials
Peptone from meat, 250 g
CAS-nummer:  
Discontinued
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
  • Suitable as nutrient for laboratory media and industrial fermentation
Specificaties
Specificaties
  • VPE:
    250 g
  • Omschrijving:
    Peptone from meat
Specificaties